Skip to Content
Merck
2,4-Dihydroxy-5-methoxybenzaldehyde 97%

2,4-Dihydroxy-5-methoxybenzaldehyde

2,4-Dihydroxy-5-methoxybenzaldehyde 97%
Empirical Formula (Hill Notation):
C8H8O4
CAS Number:
Molecular Weight:
168.15

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing