Skip to Content
Merck
2,4,6-Trimethylbenzenesulfonohydrazide 97%

2,4,6-Trimethylbenzenesulfonohydrazide

2,4,6-Trimethylbenzenesulfonohydrazide 97%
Synonym(s):
2-Mesitylenesulfonohydrazide
Linear Formula:
(CH3)3C6H2SO2NHNH2
CAS Number:
Molecular Weight:
214.28
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing