![2-(3-Chloropropoxy)tetrahydro-2H-pyran 97%](/deepweb/assets/sigmaaldrich/product/structures/134/848/8f85b813-b196-4392-a243-b35a49976e22/640/8f85b813-b196-4392-a243-b35a49976e22.png)
2-(3-Chloropropoxy)tetrahydro-2H-pyran
![2-(3-Chloropropoxy)tetrahydro-2H-pyran 97%](/deepweb/assets/sigmaaldrich/product/structures/134/848/8f85b813-b196-4392-a243-b35a49976e22/640/8f85b813-b196-4392-a243-b35a49976e22.png)
Empirical Formula (Hill Notation):
C8H15ClO2
CAS Number:
Molecular Weight:
178.66
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing