Skip to Content
Merck
2,3,4,6-Tetra-O-benzyl-D-galactose ≥98%

2,3,4,6-Tetra-O-benzyl-D-galactose

2,3,4,6-Tetra-O-benzyl-D-galactose ≥98%
Empirical Formula (Hill Notation):
C34H36O6
CAS Number:
Molecular Weight:
540.65

Products

Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Optical Isomer
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing