Skip to Content
Merck
2-(2-Bromoethyl)-1,3-dioxane 98%

2-(2-Bromoethyl)-1,3-dioxane

2-(2-Bromoethyl)-1,3-dioxane 98%
Synonym(s):
3-Bromopropionaldehyde trimethylene acetal
Empirical Formula (Hill Notation):
C6H11BrO2
CAS Number:
Molecular Weight:
195.05
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing