
2-(2-Bromoethoxy)tetrahydro-2H-pyran

Empirical Formula (Hill Notation):
C7H13BrO2
CAS Number:
Molecular Weight:
209.08
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing