Skip to Content
Merck
2-(1,3-Dioxan-2-yl)ethyltriphenylphosphonium bromide 98%

2-(1,3-Dioxan-2-yl)ethyltriphenylphosphonium bromide

2-(1,3-Dioxan-2-yl)ethyltriphenylphosphonium bromide 98%
Empirical Formula (Hill Notation):
C24H26BrO2P
CAS Number:
Molecular Weight:
457.34
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing