Skip to Content
Merck
1-Trimethylsilyl-1-hexyne 99%

1-Trimethylsilyl-1-hexyne

1-Trimethylsilyl-1-hexyne 99%
Linear Formula:
CH3(CH2)3C≡CSi(CH3)3
CAS Number:
Molecular Weight:
154.32

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing