Skip to Content
Merck
(1R)-(−)-10-Camphorsulfonyl chloride 97%

(1R)-(−)-10-Camphorsulfonyl chloride

(1R)-(−)-10-Camphorsulfonyl chloride 97%
Synonym(s):
(−)-Camphor-10-sulfonyl chloride, (1R)-Camphor-10-sulfonic acid chloride
Empirical Formula (Hill Notation):
C10H15ClO3S
CAS Number:
Molecular Weight:
250.74

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing