
1-(p-Toluenesulfonyl)azetidine

Synonym(s):
1-Tosylazetidine
Empirical Formula (Hill Notation):
C10H13NO2S
CAS Number:
Molecular Weight:
211.28
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing