Skip to Content
Merck
1-Piperazinepropanol 97%

1-Piperazinepropanol

1-Piperazinepropanol 97%
Synonym(s):
1-(3-Hydroxy-1-propyl)piperazine, 1-(3-Hydroxypropyl)-4-piperazine, 1-(3-Hydroxypropyl)piperazine, 3-(1-Piperazinyl)-1-propanol, 3-(1-Piperazinyl)propanol, 4-(3-Hydroxypropyl)piperazine, N-(3-Hydroxypropyl)piperazine
Empirical Formula (Hill Notation):
C7H16N2O
CAS Number:
Molecular Weight:
144.21
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing