Skip to Content
Merck
1-Methyl-1H-imidazole-5-carboxylic acid

1-Methyl-1H-imidazole-5-carboxylic acid

1-Methyl-1H-imidazole-5-carboxylic acid
Synonym(s):
1-Methylimidazole-5-carboxylic acid
Empirical Formula (Hill Notation):
C5H6N2O2
CAS Number:
Molecular Weight:
126.11

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing