Skip to Content
Merck
1-Fluoropyridinium tetrafluoroborate ≥95%

1-Fluoropyridinium tetrafluoroborate

1-Fluoropyridinium tetrafluoroborate ≥95%
Empirical Formula (Hill Notation):
C5H5BF5N
CAS Number:
Molecular Weight:
184.90

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing