Skip to Content
Merck
1-Ethyl-4-ethynylbenzene 98%

1-Ethyl-4-ethynylbenzene

1-Ethyl-4-ethynylbenzene 98%
Linear Formula:
CH3CH2C6H4C≡CH
CAS Number:
Molecular Weight:
130.19

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing