Skip to Content
Merck
1-Cyclopentene-1,2-dicarboxylic anhydride 96%

1-Cyclopentene-1,2-dicarboxylic anhydride

1-Cyclopentene-1,2-dicarboxylic anhydride 96%
Empirical Formula (Hill Notation):
C7H6O3
CAS Number:
Molecular Weight:
138.12

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Monomer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing