Skip to Content
Merck
1-(cis-3-Chloroallyl)-3,5,7-triaza-1-azoniaadamantane chloride ≥97%

1-(cis-3-Chloroallyl)-3,5,7-triaza-1-azoniaadamantane chloride

1-(cis-3-Chloroallyl)-3,5,7-triaza-1-azoniaadamantane chloride ≥97%
Empirical Formula (Hill Notation):
C9H16Cl2N4
CAS Number:
Molecular Weight:
251.16

Products

Brand
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing