Skip to Content
Merck
1-Chloro-3-pentanone technical grade, 85%

1-Chloro-3-pentanone

1-Chloro-3-pentanone technical grade, 85%
Linear Formula:
CH3CH2COCH2CH2Cl
CAS Number:
Molecular Weight:
120.58
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing