Skip to Content
Merck
1-Butyl-1-methylpyrrolidinium tetrafluoroborate 98%, H2O <500 ppm

1-Butyl-1-methylpyrrolidinium tetrafluoroborate

1-Butyl-1-methylpyrrolidinium tetrafluoroborate 98%, H2O <500 ppm
Empirical Formula (Hill Notation):
C9H20BF4N
CAS Number:
Molecular Weight:
229.07

Products

Brand
Formula Weight
Physical Form
Purity
Chemical Composition
Application
Markush Class
Markush Group