Skip to Content
Merck
1-(Boc-amino)cyclopropanecarboxylic acid ≥98.0% (TLC)

1-(Boc-amino)cyclopropanecarboxylic acid

1-(Boc-amino)cyclopropanecarboxylic acid ≥98.0% (TLC)
Empirical Formula (Hill Notation):
C9H15NO4
CAS Number:
Molecular Weight:
201.22
EC Number:

Products

Brand
Formula Weight
Physical Form
Purity
Application
Markush Class
Markush Group
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing