Skip to Content
Merck
1-Bis(4-fluorophenyl)methyl piperazine 97%

1-Bis(4-fluorophenyl)methyl piperazine

1-Bis(4-fluorophenyl)methyl piperazine 97%
Synonym(s):
1-(4,4′-Difluorobenzhydryl)piperazine
Empirical Formula (Hill Notation):
C17H18F2N2
CAS Number:
Molecular Weight:
288.34
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing