
1-Benzylindole-3-carboxylic acid

Empirical Formula (Hill Notation):
C16H13NO2
CAS Number:
Molecular Weight:
251.28
Products
Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing