Skip to Content
Merck
1-Aminopyrrolidine hydrochloride technical grade

1-Aminopyrrolidine hydrochloride

1-Aminopyrrolidine hydrochloride technical grade
Empirical Formula (Hill Notation):
C4H10N2 · HCl
CAS Number:
Molecular Weight:
122.60
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing