1-Acetyl-6-aminoindoline
Synonym(s):
1-(6-Amino-2,3-dihydro-indol-1-yl)ethanone, 1-Acetyl-6-amino-2,3-dihydroindole, 6-Amino-N-acetylindoline, NSC 87623
Empirical Formula (Hill Notation):
C10H12N2O
CAS Number:
Molecular Weight:
176.22
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing