Skip to Content
Merck
1,9-Nonanedithiol 99%

1,9-Nonanedithiol

1,9-Nonanedithiol 99%
Synonym(s):
1,9-Dimercaptononane
Linear Formula:
HS(CH2)9SH
CAS Number:
Molecular Weight:
192.39
EC Number:

Products

Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing