Skip to Content
Merck
1-(4-Chlorobenzyl)piperazine 98%

1-(4-Chlorobenzyl)piperazine

1-(4-Chlorobenzyl)piperazine 98%
Empirical Formula (Hill Notation):
C11H15ClN2
CAS Number:
Molecular Weight:
210.70

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing