Skip to Content
Merck
1,4,8,12-Tetraazacyclopentadecane 97%

1,4,8,12-Tetraazacyclopentadecane

1,4,8,12-Tetraazacyclopentadecane 97%
Empirical Formula (Hill Notation):
C11H26N4
CAS Number:
Molecular Weight:
214.35

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Monomer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing