Skip to Content
Merck
1,3-Diethyl-1,1,3,3-tetramethyldisilazane >98.0%

1,3-Diethyl-1,1,3,3-tetramethyldisilazane

1,3-Diethyl-1,1,3,3-tetramethyldisilazane >98.0%
Empirical Formula (Hill Notation):
C8H23NSi2
CAS Number:
Molecular Weight:
189.45

Products

Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing