Skip to Content
Merck
1-(2-Thienyl)-1-propanone 98%

1-(2-Thienyl)-1-propanone

1-(2-Thienyl)-1-propanone 98%
Synonym(s):
2-Propionylthiophene
Empirical Formula (Hill Notation):
C7H8OS
CAS Number:
Molecular Weight:
140.20
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing