Skip to Content
Merck
1-(2-Pyrimidyl)piperazine dihydrochloride ≥96.0% (HPLC)

1-(2-Pyrimidyl)piperazine dihydrochloride

1-(2-Pyrimidyl)piperazine dihydrochloride ≥96.0% (HPLC)
Synonym(s):
2-(1-Piperazinyl)pyrimidine dihydrochloride, 2-Piperazinopyrimidine dihydrochloride
Empirical Formula (Hill Notation):
C8H12N4 · 2HCl
CAS Number:
Molecular Weight:
237.13
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing