
1,2-Didodecanoyl-rac-glycero-3-phosphocholine

Synonym(s):
4-Hydroxy-N,N,N-trimethyl-10-oxo-7-[(1-oxododecyl)oxy]-3,5,9-trioxa-4-phosphaheneicosan-1-aminium 4-oxide, inner salt, rac-1,2-Dilauroyl-glycero-3-phosphocholine, rac-DLPC, rac-Phosphatidylcholine, 1,2-dilauroyl, DL-α-Dilauroyl phosphatidylcholine, DL-β,γ-Dilauroyl-α-lecithin, PC(12:0/12:0)[rac]
Empirical Formula (Hill Notation):
C32H64NO8P
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Functional Group
Showing 1-1 of 1