Skip to Content
Merck
1,2-Bis(trimethylsiloxy)cyclobutene ≥95%

1,2-Bis(trimethylsiloxy)cyclobutene

1,2-Bis(trimethylsiloxy)cyclobutene ≥95%
Empirical Formula (Hill Notation):
C10H22O2Si2
CAS Number:
Molecular Weight:
230.45

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing