Skip to Content
Merck
1-(2-Aminoethyl)piperidine 98%

1-(2-Aminoethyl)piperidine

1-(2-Aminoethyl)piperidine 98%
Synonym(s):
2-Piperidinoethylamine
Empirical Formula (Hill Notation):
C7H16N2
CAS Number:
Molecular Weight:
128.22
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing