
1,1′-Oxalyldiimidazole

Synonym(s):
Oxalic acid diimidazolide
Empirical Formula (Hill Notation):
C8H6N4O2
CAS Number:
Molecular Weight:
190.16
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Application
Markush Class
Markush Group
Reaction Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing