Skip to Content
Merck
1-(1-Naphthyl)ethylamine 98%

1-(1-Naphthyl)ethylamine

1-(1-Naphthyl)ethylamine 98%
Synonym(s):
(±)-1-(1-Naphthyl)ethylamine, (±)-α-Methyl-1-naphthalenemethylamine
Linear Formula:
C10H7CH(CH3)NH2
CAS Number:
Molecular Weight:
171.24

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing