Skip to Content
Merck
All Photos(1)

Key Documents

SAB2501696

Sigma-Aldrich

Anti-KEAP1 antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

INrf2, KIAA0132, KLHL19, kelch-like ECH-associated protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

pig, human, canine, mouse, rat, bovine

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
indirect ELISA: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KEAP1(9817)

Immunogen

Peptide with sequence C-EVTPSQHGNRTFS, from the internal region of the protein sequence according toNP_036421.2

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in 20mM Tris (pH 7.3) and 150mM NaCl with 0.02% sodium azide and 0.5% bovine serum albumin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isabella Cattani-Cavalieri et al.
Inflammation, 42(2), 526-537 (2018-11-10)
Air pollution caused by fuel burning contributes to respiratory impairments that may lead to death. We aimed to investigate the effects of biodiesel (DB) burning in mouse lungs. DB particulate matter was collected from the exhaust pipes of a bus

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service