Skip to Content
Merck
All Photos(1)

Key Documents

DEN2506

Density Standard (25 °C)

UKAS ISO/IEC17025 and ISO 17034 certified, density: 0.934 g/mL at 25 °C

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Agency

according to ASTM® D1480

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Paragon Scientific Ltd

density

0.934 g/mL at 25 °C

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
industrial qc
personal care
petroleum

format

mixture

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Please note that the value stated on this page is nominal. All certified values for the current lots of all Paragon Density Standards can be found on the Paragon Scientific webpage

Application

This density CRM is for the calibration of instruments used to measure the density of materials that are fluids at the desired test temperature of 25 °C.

Features and Benefits

  • Manufactured and verified in an ISO 17025 and ISO 17034 accredited laboratory, as per UKAS
  • Carries an associated uncertainty of +/- 0.01%
  • Certified density value following ASTM D1480
  • Provided in a tamper-evident packaging to assure safe delivery
  • Marked holographically to ensure authenticity
  • Compliant to ASTM and IP test method protocols
  • Accompanied with a certificate of analysis (CoA) having information on expiry date and storage conditions for an effective audit

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

57.2 °F - closed cup

Flash Point(C)

14.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service