APO455834321
Apollo Scientific Ltd
5-Fluoro-2-methoxypyridine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
mp
<60
Iupac Name
5-fluoro-2-methoxypyridine
Inchi Code
1S/C6H6FNO/c1-9-6-3-2-5(7)4-8-6/h2-4H,1H3
InChI key
GYGABXVZIHADCT-UHFFFAOYSA-N
storage temp.
Store at -20C
Purity
97%
Country of Origin
GB
Shipping Temperature
Shipped with Ice Packs
Physical Form
Liquid
Product Link
https://store.apolloscientific.co.uk/product/5-fluoro-2-methoxypyridine
Synonym
5-Fluoropyridin-2-yl methyl ether
Flash Point
137-140
Melting Point
<60
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
https://store.apolloscientific.co.uk/product/5-fluoro-2-methoxypyridine
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service