Skip to Content
Merck
All Photos(1)

Key Documents

150231

Sigma-Aldrich

4-Chloroquinaldine

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8ClN
CAS Number:
Molecular Weight:
177.63
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

99%

form

liquid

refractive index

n20/D 1.622 (lit.)

bp

269-270 °C (lit.)

density

0.881 g/mL at 25 °C (lit.)

functional group

chloro

SMILES string

Cc1cc(Cl)c2ccccc2n1

InChI

1S/C10H8ClN/c1-7-6-9(11)8-4-2-3-5-10(8)12-7/h2-6H,1H3

InChI key

HQAIROMRVBVWSK-UHFFFAOYSA-N

Application

4-Chloroquinaldine was used in the preparation of 4-phenyl-hydrazinoquinaldine[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

208. The reaction between phenylhydrazine and 4-chloroquinoline derivatives, and the preparation of the corresponding 4-benzeneazo-and 4-amino-compounds.
Backeberg OG.
Journal of the Chemical Society, 1083-1087 (1938)
K N Vennila et al.
Bioorganic chemistry, 81, 184-190 (2018-08-24)
The induced fit docking of anilino quinoline scaffold results in the required hydrogen bonding interactions with amino acid residues in the orthosteric site of 3 Phosphoinositide dependent kinase (PDK1). The rational design of 4-substituted amino quinolines is carried out and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service