MilliporeSigma
HPLC Analysis of Lactic Acid Enantiomers on Astec® CLC-L and CLC-D
HPLC Analysis of Lactic Acid Enantiomers on Astec® CLC-L and CLC-D application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Astec® CLC-D Chiral HPLC Column
5 μm particle size, L × I.D. 15 cm × 4.6 mm
Astec® CLC-L Chiral HPLC Column
5 μm particle size, L × I.D. 15 cm × 4.6 mm
standard
Product No.
Description
Pricing
D-(−)-Lactic acid
≥90%
L-(+)-Lactic acid silver salt
~90%
DL-Lactic acid
85 % (w/w), syrup
Lactic acid
meets USP testing specifications
L-(+)-Lactic acid
≥98%
L-(+)-Lactic acid
BioXtra, ≥98% (titration)
L-(+)-Lactic acid solution
30% in H2O (by weight)
L-(+)-Lactic acid solution
meets analytical specification of Ph. Eur., BP, 88-92% total acid basis
L-(+)-Lactic acid
80%
DL-Lactic acid
~90% (T)
L-(+)-Lactic acid
analytical standard
Lactic acid
Pharmaceutical Secondary Standard; Certified Reference Material
CONDITIONS
column
Astec CLC-L and CLC-D, 15 cm x 4.6 mm I.D., 5 μm particles (53023AST, 53123AST)
mobile phase
5 mM copper sulfate
flow rate
1 mL/min
column temp.
25 °C
detector
UV, 254 nm
injection
10 μL