MilliporeSigma
Obestatin prevents H
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie (2017-11-16)
Yang Liu, Yue-Xian Xing, Xin-Yuan Gao, Hong-Yu Kuang, Jing Zhang, Rong Liu
ABSTRACT
In the early stage of diabetic retinopathy, the damage of retinal ganglion cells already exists, promoting the development of the disease. The aim of this study was to investigate the protective role and the mechanisms of obestatin against H RGC-5 cells were incubated with various concentrations of obestatin for 24h before H Our data showed that H Obestatin prevents H
Sigma-Aldrich
Exendin Fragment 9-39, ≥95% (HPLC)