MilliporeSigma
Robust fabrication of thin film polyamide-TiO
Scientific reports (2018-01-18)
Behnam Khorshidi, Ishita Biswas, Tanushree Ghosh, Thomas Thundat, Mohtada Sadrzadeh
ABSTRACT
The development of nano-enabled composite materials has led to a paradigm shift in the manufacture of high-performance nanocomposite membranes with enhanced permeation, thermo-mechanical, and antibacterial properties. The major challenges to the successful incorporation of nanoparticles (NPs) to polymer films are the severe aggregation of the NPs and the weak compatibility of NPs with polymers. These two phenomena lead to the formation of non-selective voids at the interface of the polymer and NPs, which adversely affect the separation performance of the membrane. To overcome these challenges, we have developed a new method for the fabrication of robust TFN reverse osmosis membranes. This approach relies on the simultaneous synthesis and surface functionalization of TiO
Sigma-Aldrich
1,3,5-Benzenetricarbonyl trichloride, 98%