MilliporeSigma
  • Zn-mediated, Pd-catalyzed cross-couplings in water at room temperature without prior formation of organozinc reagents.
Zn-mediated, Pd-catalyzed cross-couplings in water at room temperature without prior formation of organozinc reagents.
Journal of the American Chemical Society (2009-10-16)
Arkady Krasovskiy, Christophe Duplais, Bruce H Lipshutz
ABSTRACT
Mix in water, stir. That is all that is required in this new approach to sp(3)-sp(2) cross-couplings between an alkyl iodide and an aryl bromide, both potentially bearing functionality. They react under catalysis by Pd(0) in the presence of zinc powder, aided by a nonionic amphiphile, to give the alkylated aromatic. No organic solvents and no heating; just add water.
Sigma-Aldrich
Bis(di-tert-butyl(4-dimethylaminophenyl)phosphine)dichloropalladium(II)