MilliporeSigma
  • Glycogen Synthase Kinase-3 Modulates Cbl-b and Constrains T Cell Activation.
Glycogen Synthase Kinase-3 Modulates Cbl-b and Constrains T Cell Activation.
Journal of immunology (Baltimore, Md. : 1950) (2017-11-08)
Charles W Tran, Samuel D Saibil, Thierry Le Bihan, Sara R Hamilton, Karl S Lang, Han You, Amy E Lin, Kristine M Garza, Alisha R Elford, Kelly Tai, Michael E Parsons, Kip Wigmore, Mitchell G Vainberg, Josef M Penninger, James R Woodgett, Tak W Mak, Pamela S Ohashi
ABSTRACT
The decision between T cell activation and tolerance is governed by the spatial and temporal integration of diverse molecular signals and events occurring downstream of TCR and costimulatory or coinhibitory receptor engagement. The PI3K-protein kinase B (PKB; also known as Akt) signaling pathway is a central axis in mediating proximal signaling events of TCR and CD28 engagement in T cells. Perturbation of the PI3K-PKB pathway, or the loss of negative regulators of T cell activation, such as the E3 ubiquitin ligase Cbl-b, have been reported to lead to increased susceptibility to autoimmunity. In this study, we further examined the molecular pathway linking PKB and Cbl-b in murine models. Our data show that the protein kinase GSK-3, one of the first targets identified for PKB, catalyzes two previously unreported phosphorylation events at Ser
Sigma-Aldrich
1-Azakenpaullone, ≥97% (HPLC)