MilliporeSigma
  • Influence of surface coating on the intracellular behaviour of gold nanoparticles: a fluorescence correlation spectroscopy study.
Influence of surface coating on the intracellular behaviour of gold nanoparticles: a fluorescence correlation spectroscopy study.
Nanoscale (2017-09-28)
A Silvestri, D Di Silvio, I Llarena, R A Murray, M Marelli, L Lay, L Polito, S E Moya
ABSTRACT
In the biomedical applications of nanoparticles (NPs), the proper choice of surface chemistry is a crucial aspect in their design. The nature of the coating can heavily impact the interaction of NPs with biomolecules, affect the state of aggregation, and ultimately determine their biological fate. As such, protein corona formation and the aggregation behaviour of gold NPs (Au NPs) are studied here. Au NPs are prepared with four distinct surface functionalisations, namely mercaptosuccinic acid (MSA), N-4-thiobutyroil glucosamine, HS-PEG
Sigma-Aldrich
4,4′-Dithiodibutyric acid, 95%