MilliporeSigma
  • Cesium-Induced Ionic Conduction through a Single Nanofluidic Pore Modified with Calixcrown Moieties.
Cesium-Induced Ionic Conduction through a Single Nanofluidic Pore Modified with Calixcrown Moieties.
Langmuir : the ACS journal of surfaces and colloids (2017-08-11)
Mubarak Ali, Ishtiaq Ahmed, Patricio Ramirez, Saima Nasir, Javier Cervera, Salvador Mafe, Christof M Niemeyer, Wolfgang Ensinger
ABSTRACT
We demonstrate experimentally and theoretically a nanofluidic device for the selective recognition of the cesium ion by exploiting host-guest interactions inside confined geometry. For this purpose, a host molecule, i.e., the amine-terminated p-tert-butylcalix[4]arene-crown (t-BuC[4]C-NH
Sigma-Aldrich
Pentaethylene glycol di(p-toluenesulfonate), 95%