MilliporeSigma
  • Blomia tropicalis allergen 5 (Blo t 5) T-cell epitopes and their ability to suppress the allergic immune response.
Blomia tropicalis allergen 5 (Blo t 5) T-cell epitopes and their ability to suppress the allergic immune response.
Immunology (2017-06-06)
Kenneth H Wong, Qian Zhou, Nayana Prabhu, Kazuki Furuhashi, Yen Leong Chua, Gijsbert M Grotenbreg, David M Kemeny
ABSTRACT
Blomia tropicalis is the major asthma allergen in the tropics comparable to Dermatophagoides pteronyssinus. However, little is known about the B. tropicalis epitopes recognized by T cells. Our aim was to identify the T-cell epitopes in the major B. tropicalis allergen, Blo t 5, and investigate the potential of the corresponding peptides to inhibit the allergic inflammatory lung response. C57BL/6 mice were immunized with plasmid DNA encoding Blo t 5 and T-cell epitopes identified using the interferon-γ ELISPOT assay with 15-mer overlapping peptides. C57BL/6 mice were sensitized with bone-marrow-derived dendritic cells (BMDC) pulsed with Blo t 5 allergen followed by intranasal Blo t 5 challenge. Two H-2
Sigma-Aldrich
Mouse IgE ELISA Kit