MilliporeSigma
  • M2-like macrophages induce colon cancer cell invasion via matrix metalloproteinases.
M2-like macrophages induce colon cancer cell invasion via matrix metalloproteinases.
Journal of cellular physiology (2017-01-18)
Katyayni Vinnakota, Yuan Zhang, Benson Chellakkan Selvanesan, Geriolda Topi, Tavga Salim, Janna Sand-Dejmek, Gunilla Jönsson, Anita Sjölander
ABSTRACT
The inflammatory milieu plays an important role in colon cancer development and progression. Previously, we have shown that tumor-associated macrophages (TAMs), an important component of the tumor microenvironment, are enriched in tumors compared with normal tissue and confer a poorer prognosis. In the present study, we found that matrix metallopeptidase-9 (MMP-9), which degrades extracellular matrix proteins, was increased in biopsies from colon cancer patients and in mouse xenografts with SW480 cell-derived tumors. SW480 colon cancer cells exposed to M2-like macrophage-conditioned medium (M2-medium) exhibited increased MMP-9 mRNA, protein expression and gelatinase activity. A similar effect was obtained by the addition of tumor necrosis factor-α (TNFα) and leukotriene D
Sigma-Aldrich
PP1, ≥98% (HPLC)