MilliporeSigma
  • Which intervention is better for malaria vector control: insecticide mixture long-lasting insecticidal nets or standard pyrethroid nets combined with indoor residual spraying?
Which intervention is better for malaria vector control: insecticide mixture long-lasting insecticidal nets or standard pyrethroid nets combined with indoor residual spraying?
Malaria journal (2017-08-18)
Corine Ngufor, Josias Fagbohoun, Jessica Critchley, Raphael N'Guessan, Damien Todjinou, David Malone, Martin Akogbeto, Mark Rowland
ABSTRACT
Malaria control today is threatened by widespread insecticide resistance in vector populations. The World Health Organization (WHO) recommends the use of a mixture of unrelated insecticides for indoor residual spraying (IRS) and long-lasting insecticidal nets (LNs) or as a combination of interventions for improved vector control and insecticide resistance management. Studies investigating the efficacy of these different strategies are necessary. The efficacy of Interceptor WHO resistance bioassays performed during the trial demonstrated a pyrethroid resistance frequency of >90% in the wild An. gambiae s.l. from the Cove hut site. Mortality in the control (untreated net) hut was 5%. Mortality with Interceptor The use of chlorfenapyr and alpha-cypermethrin together as a mixture on nets (Interceptor
Product Description
Supelco
Chlorfenapyr, PESTANAL®, analytical standard