MilliporeSigma
Regulation of
Development (Cambridge, England) (2017-03-31)
Qi Zhang, Jamie Zagozewski, Shaohong Cheng, Rajiv Dixit, Shunzhen Zhang, Jimmy de Melo, Xiuqian Mu, William H Klein, Nadean L Brown, Jeffrey T Wigle, Carol Schuurmans, David D Eisenstat
ABSTRACT
Regulated retinal ganglion cell (RGC) differentiation and axonal guidance is required for a functional visual system. Homeodomain and basic helix-loop-helix transcription factors are required for retinogenesis, as well as patterning, differentiation and maintenance of specific retinal cell types. We hypothesized that
Sigma-Aldrich
Cell dissociation sieve - tissue grinder kit, tissue grinder homogenizer kit, stainless steel, autoclavable, 1/kit