MilliporeSigma
  • Sulfur-doped graphene derived from cycled lithium-sulfur batteries as a metal-free electrocatalyst for the oxygen reduction reaction.
Sulfur-doped graphene derived from cycled lithium-sulfur batteries as a metal-free electrocatalyst for the oxygen reduction reaction.
Angewandte Chemie (International ed. in English) (2014-12-09)
Zhaoling Ma, Shuo Dou, Anli Shen, Li Tao, Liming Dai, Shuangyin Wang
ABSTRACT
Heteroatom-doped carbon materials have been extensively investigated as metal-free electrocatalysts to replace commercial Pt/C catalysts in oxygen reduction reactions in fuel cells and Li-air batteries. However, the synthesis of such materials usually involves high temperature or complicated equipment. Graphene-based sulfur composites have been recently developed to prolong the cycling life of Li-S batteries, one of the most attractive energy-storage devices. Given the high cost of graphene, there is significant demand to recycle and reuse graphene from Li-S batteries. Herein, we report a green and cost-effective method to prepare sulfur-doped graphene, achieved by the continuous charge/discharge cycling of graphene-sulfur composites in Li-S batteries. This material was used as a metal-free electrocatalyst for the oxygen reduction reaction and shows better electrocatalytic activity than pristine graphene and better methanol tolerance durability than Pt/C.
Sigma-Aldrich
Sulfur-doped graphene, avg. no. of layers, 1 ‑ 5